missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dr1-associated corepressor Protein
A DNA sequence encoding the Homo sapiens (Human) Dr1-associated corepressor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8494-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_006433.2 | |
Dr1-associated corepressor Protein | |
Research Use Only | |
DRAP1 | |
Recombinant Protein | |
E. coli | |
KKKKYNARFP PARIKKIMQT DEEIGKVAAA VPVIISRALE LFLESLLKKA CQVTQSRNAK TMTTSHLKQC IELEQQFDFL KDLVASVPDM QGDGEDNHMD GDKGARRGRK PGSGGRKNGG MGTKSKDKKL SGTDSEQEDE SEDTDTDGEE ETSQPPPQAS HPSAHFQSPP TPFLPFASTL PLPPAPPGPS APDEE | |
Greater than 90% as determined by SDS-PAGE. |
10589 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |