missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DUSP23 Protein

A cDNA sequence encoding the DUSP23 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10578-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944308

  • 1545.00 SEK / 2µg
Estimated Shipment: 15-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

54935
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 200 units/mg, and is defined as the amount of enzyme that hydrolyzes 1.0 nmole of p-nitrophenyl phosphate (pNPP) per minute at pH 7.5 at 37C.
Human
His
DUSP23 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 10% glycerol and 100mM NaCl.
DUSP23 Protein
Research Use Only
DUSP23
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGVQPPNFSW VLPGRLAGLA LPRLPAHYQF LLDLGVRHLV SLTERGPPHS DSCPGLTLHR LRIPDFCPPA PDQIDRFVQI VDEANARGEA VGVHCALGFG RTGTMLACYL VKERGLAAGD AIAEIRRLRP GSIETYEQEK AVFQFYQRTK
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers