missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase MYLIP Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase MYLIP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7993-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037394.2 | |
E3 ubiquitin-protein ligase MYLIP Protein | |
Research Use Only | |
MYLIP | |
Recombinant Protein | |
E. coli | |
MLCYVTRPDA VLMEVEVEAK ANGEDCLNQV CRRLGIIEVD YFGLQFTGSK GESLWLNLRN RISQQMDGLA PYRLKLRVKF FVEPHLILQE QTRHIFFLHI KEALLAGHLL CSPEQAVELS ALLAQTKFGD YNQNTAKYNY EELCAKELSS ATLNSIVAKH KELEGTSQAS AEYQVLQIVS AMENYGIEWH SVRDSEGQKL LIGVGPEGIS ICKDDFSPIN RIAYPVVQMA TQSGKNVYLT VTKESGNSIV LLFKMISTRA ASGLYRAITE THAFYRCDTV TSAVMMQYSR DLKGHLASLF LNENINLGKK YVFDIKRTSK EVYDHARRAL YNAGVVDLVS RNNQSPSHSP LKSSESSMNC SSCEGLSCQQ TRVLQEKLRK LKEAMLCMVC CEEEINSTFC PCGHTVCCES CAAQLQSCPV CRSRVEHVQH VYLPTHTSLL NLTVI | |
Greater than 90% as determined by SDS-PAGE. |
29116 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |