missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase NRDP1 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase NRDP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8174-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001229755.1 | |
E3 ubiquitin-protein ligase NRDP1 Protein | |
Research Use Only | |
RNF41 | |
Recombinant Protein | |
E. coli | |
MGYDVTRFQG DVDEDLICPI CSGVLEEPVQ APHCEHAFCN ACITQWFSQQ QTCPVDRSVV TVAHLRPVPR IMRNMLSKLQ IACDNAVFGC SAVVRLDNLM SHLSDCEHNP KRPVTCEQGC GLEMPKDELP NHNCIKHLRS VVQQQQTRIA ELEKTSAEHK HQLAEQKRDI QLLKAYMRAI RSVNPNLQNL EETIEYNEIL EWVNSLQPAR VTRWGGMIST PDAVLQAVIK RSLVESGCPA SIVNELIENA HERSWPQGLA TLETRQMNRR YYENYVAKRI PGKQAVVVMA CENQHMGDDM VQEPGLVMIF AHGVEEI | |
Greater than 80% as determined by SDS-PAGE. |
10193 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |