missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase TRIM11 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase TRIM11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8004-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_660215.1 | |
E3 ubiquitin-protein ligase TRIM11 Protein | |
Research Use Only | |
TRIM11 | |
Recombinant Protein | |
E. coli | |
MELRTVCRVP GLVETLRRFR GDVTLDPDTA NPELILSEDR RSVQRGDLRQ ALPDSPERFD PGPCVLGQER FTSGRHYWEV EVGDRTSWAL GVCRENVNRK EKGELSAGNG FWILVFLGSY YNSSERALAP LRDPPRRVGI FLDYEAGHLS FYSATDGSLL FIFPEIPFSG TLRPLFSPLS SSPTPMTICR PKGGSGDTLA PQ | |
Greater than 90% as determined by SDS-PAGE. |
81559 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |