missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase TRIM21 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase TRIM21, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9447-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003132.2 | |
E3 ubiquitin-protein ligase TRIM21 Protein | |
Research Use Only | |
TRIM21 | |
Recombinant Protein | |
Yeast | |
MASAARLTMM WEEVTCPICL DPFVEPVSIE CGHSFCQECI SQVGKGGGSV CPVCRQRFLL KNLRPNRQLA NMVNNLKEIS QEAREGTQGE RCAVHGERLH LFCEKDGKAL CWVCAQSRKH RDHAMVPLEE AAQEYQEKLQ VALGELRRKQ ELAEKLEVEI AIKRADWKKT VETQKSRIHA EFVQQKNFLV EEEQRQLQEL EKDEREQLRI LGEKEAKLAQ QSQALQELIS ELDRRCHSSA LELLQEVIIV LERSESWNLK DLDITSPELR SVCHVPGLKK MLRTCAVHIT LDPDTANPWL ILSEDRRQVR LGDTQQSIPG NEERFDSYPM VLGAQHFHSG KHYWEVDVTG KEAWDLGVCR DSVRRKGHFL LSSKSGFWTI WLWNKQKYEA GTYPQTPLHL QVPPCQVGIF LDYEAGMVSF YNITDHGSLI YSFSECAFTG PLRPFFSPGF NDGGKNTAPL TLCPLNIGSQ GSTDY | |
Greater than 90% as determined by SDS-PAGE. |
6737 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |