missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase TRIM69 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase TRIM69, was expressed in the hosts and tags indicated.
2430.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_001288073.1 |
---|---|
Gene ID (Entrez) | 140691 |
Name | E3 ubiquitin-protein ligase TRIM69 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16010851
|
enQuireBio™
QP7796-EC-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16030851
|
enQuireBio™
QP7796-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020851
|
enQuireBio™
QP7796-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040851
|
enQuireBio™
QP7796-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050851
|
enQuireBio™
QP7796-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060851
|
enQuireBio™
QP7796-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001288073.1 | |
E3 ubiquitin-protein ligase TRIM69 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
140691 | |
Research Use Only | |
TRIM69 | |
Recombinant Protein | |
E. coli | |
MEEELAIQQG QLETTLKELQ TLRNMQKEAI AAHKENKLHL QQHVSMEFLK LHQFLHSKEK DILTELREEG KALNEEMELN LSQLQEQCLL AKDMLVSIQA KTEQQNSFDF LKDITTLLHS LEQGMKVLAT RELISRKLNL GQYKGPIQYM VWREMQDTLC PGLSPLTLDP KTAHPNLVLS KSQTSVWHGD IKKIMPDDPE RFDSSVAVLG SRGFTSGKWY WEVEVAKKTK WTVGVVRESI IRKGSCPLTP EQGFWLLRLR NQTDLKALDL PSFSLTLTNN LDKVGIYLDY EGGQLSFYNA KTMTHIYTFS NTFMEKLYPY FCPCLNDGGE NKEPLHILHP Q | |
Greater than 80% as determined by SDS-PAGE. |