missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase TRIM69 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase TRIM69, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7796-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001288073.1 | |
E3 ubiquitin-protein ligase TRIM69 Protein | |
Research Use Only | |
TRIM69 | |
Recombinant Protein | |
E. coli | |
MEEELAIQQG QLETTLKELQ TLRNMQKEAI AAHKENKLHL QQHVSMEFLK LHQFLHSKEK DILTELREEG KALNEEMELN LSQLQEQCLL AKDMLVSIQA KTEQQNSFDF LKDITTLLHS LEQGMKVLAT RELISRKLNL GQYKGPIQYM VWREMQDTLC PGLSPLTLDP KTAHPNLVLS KSQTSVWHGD IKKIMPDDPE RFDSSVAVLG SRGFTSGKWY WEVEVAKKTK WTVGVVRESI IRKGSCPLTP EQGFWLLRLR NQTDLKALDL PSFSLTLTNN LDKVGIYLDY EGGQLSFYNA KTMTHIYTFS NTFMEKLYPY FCPCLNDGGE NKEPLHILHP Q | |
Greater than 80% as determined by SDS-PAGE. |
140691 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |