missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Early growth response protein 1
A DNA sequence encoding the Homo sapiens (Human) Early growth response protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9116-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001955.1 | |
Early growth response protein 1 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1958 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
EGR1 | |
Recombinant Protein | |
Yeast | |
SPVATSYPSP VTTSYPSPAT TSYPSPVPTS FSSPGSSTYP SPVHSGFPSP SVATTYSSVP PAFPAQVSSF PSSAVTNSFS ASTGLSDMTA TFSPRTIEIC |