missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EDNRB / Endothelin B receptor Protein
A DNA sequence encoding the Homo sapiens (Human) EDNRB / Endothelin B receptor, was expressed in the hosts and tags indicated.
8250.00 SEK - 18925.00 SEK
Specifications
Accession Number | NP_000106.1 |
---|---|
Gene ID (Entrez) | 1910 |
Name | EDNRB / Endothelin B receptor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049081
|
enQuireBio™
QP9953-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059081
|
enQuireBio™
QP9953-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039081
|
enQuireBio™
QP9953-IV-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000106.1 | |
EDNRB / Endothelin B receptor Protein | |
Not determined. | |
Full Length Transmembrane Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1910 | |
Research Use Only | |
EDNRB | |
Recombinant Protein | |
E. coli | |
MASMTGGQQM GRGSEERGFP PDRATPLLQT AEIMTPPTKT LWPKGSNASL ARSLAPAEVP KGDRTAGSPP RTISPPPCQG PIEIKETFKY INTVVSCLVF VLGIIGNSTL LRIIYKNKCM RNGPNILIAS LALGDLLHIV IDIPINVYKL LAEDWPFGAE MCKLVPFIQK ASVGITVLSL CALSIDRYRA VASWSRIKGI GVPKWTAVEI VLIWVVSVVL AVPEAIGFDI ITMDYKGSYL RICLLHPVQK TAFMQFYKTA KDWWLFSFYF CLPLAITAFF YTLMTCEMLR KKSGMQIALN DHLKQRREVA KTVFCLVLVF ALCWLPLHLS RILKLTLYNQ NDPNRCELLS FLLVLDYIGI NMASLNSCIN PIALYLVSKR FKNCFKSCLC CWCQSFEEKQ SLEEKQSCLK FKANDHGYDN FRSSNKYSSS | |
Greater than 85% as determined by SDS-PAGE. |