missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EDNRB / Endothelin B receptor Protein
A DNA sequence encoding the Homo sapiens (Human) EDNRB / Endothelin B receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9953-iv-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000106.1 | |
EDNRB / Endothelin B receptor Protein | |
50 μg | |
Not determined. | |
Full Length Transmembrane Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1910 | |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
EDNRB | |
Recombinant Protein | |
Invitro-E. coli | |
MASMTGGQQM GRGSEERGFP PDRATPLLQT AEIMTPPTKT LWPKGSNASL ARSLAPAEVP KGDRTAGSPP RTISPPPCQG PIEIKETFKY INTVVSCLVF VLGIIGNSTL LRIIYKNKCM RNGPNILIAS LALGDLLHIV IDIPINVYKL LAEDWPFGAE MCKLVPFIQK ASVGITVLSL CALSIDRYRA VASWSRIKGI GVPKWTAVEI VLIWVVSVVL AVPEAIGFDI ITMDYKGSYL RICLLHPVQK TAFMQFYKTA KDWWLFSFYF CLPLAITAFF YTLMTCEMLR KKSGMQIALN DHLKQRREVA KTVFCLVLVF ALCWLPLHLS RILKLTLYNQ NDPNRCELLS FLLVLDYIGI NMASLNSCIN PIALYLVSKR FKNCFKSCLC CWCQSFEEKQ SLEEKQSCLK FKANDHGYDN FRSSNKYSSS |