missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Endophilin-B2 Protein

A DNA sequence encoding the Homo sapiens (Human) Endophilin-B2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8092-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062621

  • 18585.00 SEK / 1mg


Estimated Shipment
09-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_001273974.1
Endophilin-B2 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
56904
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SH3GLB2
Recombinant Protein
E. coli
MDFNMKKLAS DAGIFFTRAV QFTEEKFGQA EKTELDAHFE NLLARADSTK NWTEKILRQT EVLLQPNPSA RVEEFLYEKL DRKVPSRVTN GELLAQYMAD AASELGPTTP YGKTLIKVAE AEKQLGAAER DFIHTASISF LTPLRNFLEG DWKTISKERR LLQNRRLDLD ACKARLKKAK AAEAKATTVP DFQETRPRNY ILSASASALW NDEVDKAEQE LRVAQTEFDR QAEVTRLLLE GISSTHVNHL RCLHEFVKSQ TTYYAQCYRH MLDLQKQLGR FPGTFVGTTE PASPPLSSTS PTTAAATMPV VPSVASLAPP GEASLCLEEV APPASGTRKA RVLYDYEAAD SSELALLADE LITVYSLPGM DPDWLIGERG NKKGKVPVTY LELLS
Videos
SDS
Documents