missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Endoplasmic reticulum resident protein 29 Protein

A DNA sequence encoding the Homo sapiens (Human) Endoplasmic reticulum resident protein 29, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8504-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16094991

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001029197.1
Endoplasmic reticulum resident protein 29 Protein
Research Use Only
C12orf8
Recombinant Protein
E. coli
PLDTVTFYKV IPKSKFVLVK FDTQYPYGEK QDEFKRLAEN SASSDDLLVA EVGISDYGDK LNMELSEKYK LDKESYPVFY LFRDGDFENP VPYTGAVKVG AIQRWLKGQG VYLGMPGCLP VYDALAGEFI RASGVEARQA LLKQGQDNLS SVKETQKKWA EQYLKIMGKI LDQGEDFPAS EMTRIARLIE KNKMSDGKKE ELQKSLNILT AF
Greater than 90% as determined by SDS-PAGE.
10961
50 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers