missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ETFA Protein
A DNA sequence encoding the Homo sapiens (Human) ETFA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6002-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000117.1 | |
ETFA Protein | |
Research Use Only | |
ETFA | |
Recombinant Protein | |
E. coli | |
MFRAAAPGQL RRAASLLRFQ STLVIAEHAN DSLAPITLNT ITAATRLGGE VSCLVAGTKC DKVAQDLCKV AGIAKVLVAQ HDVYKGLLPE ELTPLILATQ KQFNYTHICA GASAFGKNLL PRVAAKLEVA PISDIIAIKS PDTFVRTIYA GNALCTVKCD EKVKVFSVRG TSFDAAATSG GSASSEKASS TSPVEISEWL DQKLTKSDRP ELTGAKVVVS GGRGLKSGEN FKLLYDLADQ LHAAVGASRA AVDAGFVPND MQVGQTGKIV APELYIAVGI SGAIQHLAGM KDSKTIVAIN KDPEAPIFQV ADYGIVADLF KVVPEMTEIL KKK | |
Greater than 80% as determined by SDS-PAGE. |
2108 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |