missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Eukaryotic translation initiation factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Eukaryotic translation initiation factor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8492-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005792.1 | |
Eukaryotic translation initiation factor 1 Protein | |
Research Use Only | |
SUI1 | |
Recombinant Protein | |
E. coli | |
MSAIQNLHSF DPFADASKGD DLLPAGTEDY IHIRIQQRNG RKTLTTVQGI ADDYDKKKLV KAFKKKFACN GTVIEHPEYG EVIQLQGDQR KNICQFLVEI GLAKDDQLKV HGF | |
Greater than 90% as determined by SDS-PAGE. |
10209 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |