missing translation for 'onlineSavingsMsg'
Läs mer

enQuireBio™ Recombinant Human Fc receptor-like protein 1

Produktkod. 16070251
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 16070251

Brand: enQuireBio™ QP7695ye50ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

A DNA sequence encoding the Homo sapiens (Human) Fc receptor-like protein 6, was expressed in the hosts and tags indicated.

Specifications

Tillträdesnummer NP_001004310.2
Gen-ID (Entrez) 343413
Namn Fc receptor-like protein 1
Kvantitet 50 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol FCRL6
Protein Längd Extracellular Domain
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art Yeast
Protein Tag His
Sekvens LYLQAWPNPV FEGDALTLRC QGWKNTPLSQ VKFYRDGKFL HFSKENQTLS MGAATVQSRG QYSCSGQVMY IPQTFTQTSE TAMVQVQELF PPPVLSAIPS PEPREGSLVT LRCQTKLHPL RSALRLLFSF HKDGHTLQDR GPHPELCIPG AKEGDSGLYW CEVAPEGGQV QKQSPQLEVR VQAPVSRPVL TLHHGPADPA VGDMVQLLCE AQRGSPPILY SFYLDEKIVG NHSAPCGGTT SLLFPVKSEQ DAGNYSCEAE NSVSRERSEP KKLSLKGSQV LFTPASNW
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.