missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fibroleukin Protein

A DNA sequence encoding the Homo sapiens (Human) Fibroleukin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7571-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16059470

  • 6685.00 SEK / 200µg


Estimated Shipment
12-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_006673.1
Fibroleukin Protein
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
10875
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FGL2
Recombinant Protein
E. coli
NNETEEIKDE RAKDVCPVRL ESRGKCEEAG ECPYQVSLPP LTIQLPKQFS RIEEVFKEVQ NLKEIVNSLK KSCQDCKLQA DDNGDPGRNG LLLPSTGAPG EVGDNRVREL ESEVNKLSSE LKNAKEEINV LHGRLEKLNL VNMNNIENYV DSKVANLTFV VNSLDGKCSK CPSQEQIQSR PVQHLIYKDC SDYYAIGKRS SETYRVTPDP KNSSFEVYCD METMGGGWTV LQARLDGSTN FTRTWQDYKA GFGNLRREFW LGNDKIHLLT KSKEMILRID LEDFNGVELY ALYDQFYVAN EFLKYRLHVG NYNGTAGDAL RFNKHYNHDL KFFTTPDKDN DRYPSGNCGL YYSSGWWFDA CLSANLNGKY YHQKYRGVRN GIFWGTWPGV SEAHPGGYKS SFKEAKMMIR PKHFKP
Videos
SDS
Documents