missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ficolin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ficolin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9759-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004099.2 | |
Ficolin-2 Protein | |
Research Use Only | |
FCN2 | |
Recombinant Protein | |
Yeast | |
LQAADTCPEV KMVGLEGSDK LTILRGCPGL PGAPGPKGEA GTNGKRGERG PPGPPGKAGP PGPNGAPGEP QPCLTGPRTC KDLLDRGHFL SGWHTIYLPD CRPLTVLCDM DTDGGGWTVF QRRVDGSVDF YRDWATYKQG FGSRLGEFWL GNDNIHALTA QGTSELRVDL VDFEDNYQFA KYRSFKVADE AEKYNLVLGA FVEGSAGDSL TFHNNQSFST KDQDNDLNTG NCAVMFQGAW WYKNCHVSNL NGRYLRGTHG SFANGINWKS GKGYNYSYKV SEMKVRPA | |
Greater than 90% as determined by SDS-PAGE. |
2220 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |