missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fizzy-related protein homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Fizzy-related protein homolog, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001129669.1 |
---|---|
Gene ID (Entrez) | 51343 |
Name | Fizzy-related protein homolog Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023601
|
enQuireBio™
QP8256-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043601
|
enQuireBio™
QP8256-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033601
|
enQuireBio™
QP8256-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053601
|
enQuireBio™
QP8256-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063601
|
enQuireBio™
QP8256-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073601
|
enQuireBio™
QP8256-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001129669.1 | |
Fizzy-related protein homolog Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
51343 | |
Research Use Only | |
FZR1 | |
Recombinant Protein | |
E. coli | |
MDQDYERRLL RQIVIQNENT MPRVTEMRRT LTPASSPVSS PSKHGDRFIP SRAGANWSVN FHRINENEKS PSQNRKAKDA TSDNGKDGLA YSALLKNELL GAGIEKVQDP QTEDRRLQPS TPEKKGLFTY SLSTKRSSPD DGNDVSPYSL SPVSNKSQKL LRSPRKPTRK ISKIPFKVLD APELQDDFYL NLVDWSSLNV LSVGLGTCVY LWSACTSQVT RLCDLSVEGD SVTSVGWSER GNLVAVGTHK GFVQIWDAAA GKKLSMLEGH TARVGALAWN AEQLSSGSRD RMILQRDIRT PPLQSERRLQ GHRQEVCGLK WSTDHQLLAS GGNDNKLLVW NHSSLSPVQQ YTEHLAAVKA IAWSPHQHGL LASGGGTADR CIRFWNTLTG QPLQCIDTGS QVCNLAWSKH ANELVSTHGY SQNQILVWKY PSLTQVAKLT GHSYRVLYLA MSPDGEAIVT GAGDETLRFW NVFSKTRSTK ESVSVLNLFT RIR | |
Greater than 90% as determined by SDS-PAGE. |