missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fusion glycoprotein F0 Protein
A DNA sequence encoding the Homo sapiens (Human) Fusion glycoprotein F0, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9634-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
Fusion glycoprotein F0 Protein | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
F | |
Recombinant Protein | |
Yeast | |
NITEEFYQST CSAVSKGYLS ALRTGWYTSV ITIELSNIKE NKCNGTDAKV KLIKQELDKY KNAVTELQLL MQSTPPTNNR ARRELPRFMN YTLNNAKKTN VTLSKKRKRR FLGFLLGVGS AIASGVAVSK VLHLEGEVNK IKSALLSTNK AVVSLSNGVS VLTSKVLDLK NYIDKQLLPI VNKQSCSISN IETVIEFQQK NNRLLEITRE FSVNAGVTTP VSTYMLTNSE LLSLINDMPI TNDQKKLMSN NVQIVRQQSY SIMSIIKEEV LAYVVQLPLY GVIDTPCWKL HTSPLCTTNT KEGSNICLTR TDRGWYCDNA GSVSFFPQAE TCKVQSNRVF CDTMNSLTLP SEINLCNVDI FNPKYDCKIM TSKTDVSSSV ITSLGAIVSC YGKTKCTASN KNRGIIKTFS NGCDYVSNKG MDTVSVGNTL YYVNKQEGKS LYVKGEPIIN FYDPLVFPSD EFDASISQVN EKINQSLAFI RKSDELLHNV NAGKSTTNIM ITT |