missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human G-protein coupled receptor 161 Protein
A DNA sequence encoding the Homo sapiens (Human) G-protein coupled receptor 161, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8467-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001254538.1 | |
G-protein coupled receptor 161 Protein | |
Research Use Only | |
GPR161 | |
Recombinant Protein | |
E. coli | |
MSLNSSLSCR KELSNLTEEE GGEGGVII | |
Greater than 90% as determined by SDS-PAGE. |
23432 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |