missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Galectin-9 Protein

Product Code. 16052290
Click to view available options
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16052290

Brand: enQuireBio™ QP6300ec200ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Galectin-9, was expressed in the hosts and tags indicated.

Specifications

Tillträdesnummer NP_001317092.1
Gen-ID (Entrez) 3965
Namn Galectin-9 Protein
Kvantitet 200 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol LGALS9
Protein Längd Full Length of Isoform 2
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag His-GST
Sekvens MAFSGSQAPY LSPAVPFSGT IQGGLQDGLQ ITVNGTVLSS SGTRFAVNFQ TGFSGNDIAF HFNPRFEDGG YVVCNTRQNG SWGPEERKTH MPFQKGMPFD LCFLVQSSDF KVMVNGILFV QYFHRVPFHR VDTISVNGSV QLSYISFQPP GVWPANPAPI TQTVIHTVQS APGQMFSTPA IPPMMYPHPA YPMPFITTIL GGLYPSKSIL LSGTVLPSAQ RFHINLCSGN HIAFHLNPRF DENAVVRNTQ IDNSWGSEER SLPRKMPFVR GQSFSVWILC EAHCLKVAVD GQHLFEYYHR LRNLPTINRL EVGGDIQLTH VQT
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.