missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDAP2 Protein
A DNA sequence encoding the Homo sapiens (Human) GDAP2, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001129061.1 |
---|---|
Gene ID (Entrez) | 54834 |
Name | GDAP2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022641
|
enQuireBio™
QP8095-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042641
|
enQuireBio™
QP8095-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032641
|
enQuireBio™
QP8095-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052641
|
enQuireBio™
QP8095-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062641
|
enQuireBio™
QP8095-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072641
|
enQuireBio™
QP8095-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001129061.1 | |
GDAP2 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
54834 | |
Research Use Only | |
GDAP2 | |
Recombinant Protein | |
E. coli | |
MDPLGAPSQF VDVDTLPSWG DSCQDELNSS DTTAEIFQED TVRSPFLYNK DVNGKVVLWK GDVALLNCTA IVNTSNESLT DKNPVSESIF MLAGPDLKED LQKLKGCRTG EAKLTKGFNL AARFIIHTVG PKYKSRYRTA AESSLYSCYR NVLQLAKEQS MSSVGFCVIN SAKRGYPLED ATHIALRTVR RFLEIHGETI EKVVFAVSDL EEGTYQKLLP LYFPRSLKEE NRSLPYLPAD IGNAEGEPVV PERQIRISEK PGAPEDNQEE EDEGLGVDLS FIGSHAFARM EGDIDKQRKL ILQGQLSEAA LQKQHQRNYN RWLCQARSED LSDIASLKAL YQTGVDNCGR TVMVVVGRNI PVTLIDMDKA LLYFIHVMDH IAVKEYVLVY FHTLTSEYNH LDSDFLKKLY DVVDVKYKRN LKAVYFVHPT FRSKVSTWFF TTFSVSGLKD KIHHVDSLHQ LFSAISPEQI DFPPFVLEYD ARVRSTRSSP SPGMVY | |
Greater than 90% as determined by SDS-PAGE. |