missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Genome polyProtein
A DNA sequence encoding the Homo sapiens (Human) Genome polyprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7370-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
Genome polyProtein | |
Research Use Only | |
VP1 | |
Recombinant Protein | |
E. coli | |
NPVENYIDSV LNEVLVVPNI QPSTSVSSHA APALDAAETG HTSSVQPEDM IETRYVITDQ TRDETSIESF LGRSGCIAMI EFNTSSDKTE HDKIGKGFKT WKVSLQEMAQ IRRKYELFTY TRFDSEITIV TAAAAQGNDS GHIVLQFMYV PPGAPVPEKR DDYTWQSGTN ASVFWQEGQP YPRFTIPFMS IASAYYMFYD GYDGDSAASK YGSVVTNDMG TICVRIVTSN QKHDSNIVCR IYHKAKHIKA WCPRPPRAVA YQHTHSTNYI PSNGEATTQI KTRPDVFTVT NV | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |