Läs mer
enQuireBio™ Recombinant Human GHRP 2 Protein
Produktkod. 15958958
Change view
Kvantitet:
5 mg
25 mg
100 mg
Förpackningsstorlek:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15958958 5 mg 5mg
15948958 25 mg 25mg
15938958 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 15958958 Leverantör enQuireBio™ Leverantörsnummer QP119685mg
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Recombinant Protein
Specifikationer
Namn GHRP 2 Protein
Kvantitet 5 mg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag Untagged
Sekvens H-D-Ala-D-2-Nal-Ala-Trp-D-Phe-Lys-NH2
Buffert The GHRP-2 hormone was lyophilized with no additives.
Renhet eller kvalitetsklass Greater than 99.0% as determined by analysis by RP-HPLC.