missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glucagon Protein
A DNA sequence encoding the Homo sapiens (Human) Glucagon, was expressed in the hosts and tags indicated.
2005.00 SEK - 22130.00 SEK
Specifications
Accession Number | NP_002045.1 |
---|---|
Gene ID (Entrez) | 2641 |
Name | Glucagon Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025711
|
enQuireBio™
QP8639-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065711
|
enQuireBio™
QP8639-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015711
|
enQuireBio™
QP8639-YE-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045711
|
enQuireBio™
QP8639-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055711
|
enQuireBio™
QP8639-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16035711
|
enQuireBio™
QP8639-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002045.1 | |
Glucagon Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2641 | |
Research Use Only | |
GCG | |
Recombinant Protein | |
Yeast | |
HSQGTFTSDY SKYLDSRRAQ DFVQWLMNTK RNRNNIA | |
Greater than 90% as determined by SDS-PAGE. |