missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glucokinase regulatory Protein
A DNA sequence encoding the Homo sapiens (Human) Glucokinase regulatory protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7596-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001477.2 | |
Glucokinase regulatory Protein | |
Research Use Only | |
GCKR | |
Recombinant Protein | |
E. coli | |
MPGTKRFQHV IETPEPGKWE LSGYEAAVPI TEKSNPLTQD LDKADAENIV RLLGQCDAEI FQEEGQALST YQRLYSESIL TTMVQVAGKV QEVLKEPDGG LVVLSGGGTS GRMAFLMSVS FNQLMKGLGQ KPLYTYLIAG GDRSVVASRE GTEDSALHGI EELKKVAAGK KRVIVIGISV GLSAPFVAGQ MDCCMNNTAV FLPVLVGFNP VSMARNDPIE DWSSTFRQVA ERMQKMQEKQ KAFVLNPAIG PEGLSGSSRM KGGSATKILL ETLLLAAHKT VDQGIAASQR CLLEILRTFE RAHQVTYSQS PKIATLMKSV STSLEKKGHV YLVGWQTLGI IAIMDGVECI HTFGADFRDV RGFLIGDHSD MFNQKAELTN QGPQFTFSQE DFLTSILPSL TEIDTVVFIF TLDDNLTEVQ TIVEQVKEKT NHIQALAHST VGQTLPIPLK KLFPSIISIT WPLLFFEYEG NFIQKFQREL STKWVLNTVS TGAHVLLGKI LQNHMLDLRI SNSKLFWRAL AMLQRFSGQS KARCIESLLR AIHFPQPLSD DIRAAPISCH VQVAHEKEQV IPIALLSLLF RCSITEAQAH LAAAPSVCEA VRSALAGPGQ KRTADPLEIL EPDVQ | |
Greater than 80% as determined by SDS-PAGE. |
2646 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |