Läs mer
enQuireBio™ Recombinant Human Glutamate receptor 3 Protein
Produktkod. p-7152804
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16080950
Brand: enQuireBio™ QP6120ye10ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Homo sapiens (Human) Glutamate receptor 3, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_000819.3
Gen-ID (Entrez) 2892
Namn Glutamate receptor 3 Protein
Kvantitet 10 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol GRIA3
Protein Längd Partial (see sequence information for more details).
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art Yeast
Protein Tag His
Sekvens SLLGHYKWEK FVYLYDTERG FSILQAIMEA AVQNNWQVTA RSVGNIKDVQ EFRRIIEEMD RRQEKRYLID CEVERINTIL EQVVILGKHS RGYHYMLANL
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.