missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutaredoxin-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutaredoxin-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6094-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001186797.1 | |
Glutaredoxin-3 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10539 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
GLRX3 | |
Recombinant Protein | |
E. coli | |
AAGAAEAAVA AVEEVGSAGQ FEELLRLKAK SLLVVHFWAP WAPQCAQMNE VMAELAKELP QVSFVKLEAE GVPEVSEKYE ISSVPTFLFF KNSQKIDRLD GAHAPELTKK VQRHASSGSF LPSANEHLKE DLNLRLKKLT HAAPCMLFMK GTPQEPRCGF SKQMVEILHK HNIQFSSFDI FSDEEVRQGL KAYSSWPTYP QLYVSGELIG GLDIIKELEA SEELDTICPK APKLEERLKV LTNKASVMLF MKGNKQEAKC GFSKQILEIL NSTGVEYETF DILEDEEVRQ GLKAYSNWPT YPQLYVKGEL VGGLDIVKEL KENGELLPIL RGEN |