missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutaredoxin / GRX1 / GLRX Protein
A DNA sequence encoding the Homo sapiens (Human) Glutaredoxin / GRX1 / GLRX, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6093-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001112362.1 | |
Glutaredoxin / GRX1 / GLRX Protein | |
Research Use Only | |
GLRX | |
Recombinant Protein | |
E. coli | |
MAQEFVNCKI QPGKVVVFIK PTCPYCRRAQ EILSQLPIKQ GLLEFVDITA TNHTNEIQDY LQQLTGARTV PRVFIGKDCI GGCSDLVSLQ QSGELLTRLK QIGALQ | |
Greater than 90% as determined by SDS-PAGE. |
2745 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |