missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutathione S-transferase kappa 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutathione S-transferase kappa 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6125-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001137151.1 | |
Glutathione S-transferase kappa 1 Protein | |
Research Use Only | |
GSTK1 | |
Recombinant Protein | |
E. coli | |
GPLPRTVELF YDVLSPYSWL GFEILCRYQN IWNINLQLRP SLITGIMKDS GNKPPGLLPR KGLYMANDLK LLRHHLQIPI HFPKDFLSVM LEKGSLSAMR FLTAVNLEHP EMLEKASREL WMRVWSRNED ITEPQSILAA AEKAGMSAEQ AQGLLEKIAT PKVKNQLKET TEAACRYGAF GLPITVAHVD GQTHMLFGSD RMELLAHLLG EKWMGPIPPA VNARL | |
Greater than 80% as determined by SDS-PAGE. |
373156 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |