missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutathione S-transferase kappa 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutathione S-transferase kappa 1, was expressed in the hosts and tags indicated.
1685.00 SEK - 19050.00 SEK
Specifications
Accession Number | NP_001137151.1 |
---|---|
Gene ID (Entrez) | 373156 |
Name | Glutathione S-transferase kappa 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16040990
|
enQuireBio™
QP6125-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080990
|
enQuireBio™
QP6125-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030990
|
enQuireBio™
QP6125-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060990
|
enQuireBio™
QP6125-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070990
|
enQuireBio™
QP6125-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050990
|
enQuireBio™
QP6125-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001137151.1 | |
Glutathione S-transferase kappa 1 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
373156 | |
Research Use Only | |
GSTK1 | |
Recombinant Protein | |
E. coli | |
GPLPRTVELF YDVLSPYSWL GFEILCRYQN IWNINLQLRP SLITGIMKDS GNKPPGLLPR KGLYMANDLK LLRHHLQIPI HFPKDFLSVM LEKGSLSAMR FLTAVNLEHP EMLEKASREL WMRVWSRNED ITEPQSILAA AEKAGMSAEQ AQGLLEKIAT PKVKNQLKET TEAACRYGAF GLPITVAHVD GQTHMLFGSD RMELLAHLLG EKWMGPIPPA VNARL | |
Greater than 80% as determined by SDS-PAGE. |