missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glycogen phosphorylase, brain form Protein

A DNA sequence encoding the Homo sapiens (Human) Glycogen phosphorylase, brain form, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8681-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065931

  • 26990.00 SEK / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002853.2
Glycogen phosphorylase, brain form Protein
Research Use Only
PYGB
Recombinant Protein
E. coli
TDSEKRKQIS VRGLAGLGDV AEVRKSFNRH LHFTLVKDRN VATPRDYFFA LAHTVRDHLV GRWIRTQQHY YERDPKRIYY LSLEFYMGRT LQNTMVNLGL QNACDEAIYQ LGLDLEELEE IEEDAGLGNG GLGRLAACFL DSMATLGLAA YGYGIRYEFG IFNQKIVNGW QVEEADDWLR YGNPWEKARP EYMLPVHFYG RVEHTPDGVK WLDTQVVLAM PYDTPVPGYK NNTVNTMRLW SAKAPNDFKL QDFNVGDYIE AVLDRNLAEN ISRVLYPNDN FFEGKELRLK QEYFVVAATL QDIIRRFKSS KFGCRDPVRT CFETFPDKVA IQLNDTHPAL SIPELMRILV DVEKVDWDKA WEITKKTCAY TNHTVLPEAL ERWPVSMFEK LLPRHLEIIY AINQRHLDHV AALFPGDVDR LRRMSVIEEG DCKRINMAHL CVIGSHAVNG VARIHSEIVK QSVFKDFYEL EPEKFQNKTN GITPRRWLLL CNPGLADTIV EKIGEEFLTD LSQLKKLLPL VSDEVFIRDV AKVKQENKLK FSAFLEKEYK VKINPSSMFD VHVKRIHEYK RQLLNCLHVV TLYNRIKRDP AKAFVPRTVM IGGKAAPGYH MAKLIIKLVT SIGDVVNHDP VVGDRLKVIF LENYRVSLAE KVIPAADLSQ QISTAGTEAS GTGNMKFMLN GALTIGTMDG ANVEMAEEAG AENLFIFGLR VEDVEALDRK GYNAREYYDH LPELKQAVDQ ISSGFFSPKE PDCFKDIVNM LMHHDRFKVF ADYEAYMQCQ AQVDQLYRNP KEWTKKVIRN IACSGKFSSD RTITEYAREI WGVEPSDLQI PPPNIPRD
Greater than 90% as determined by SDS-PAGE.
5834
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers