missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNA15 Protein
A DNA sequence encoding the Homo sapiens (Human) GNA15, was expressed in the hosts and tags indicated.
2430.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_002059.3 |
---|---|
Gene ID (Entrez) | 2769 |
Name | GNA15 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16010800
|
enQuireBio™
QP6100-EC-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16030800
|
enQuireBio™
QP6100-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020800
|
enQuireBio™
QP6100-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040800
|
enQuireBio™
QP6100-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050800
|
enQuireBio™
QP6100-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060800
|
enQuireBio™
QP6100-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002059.3 | |
GNA15 Protein | |
Not determined. | |
Full Length | |
Human | |
His-Trx | |
Tris-based buffer, 50% glycerol |
2769 | |
Research Use Only | |
GNA15 | |
Recombinant Protein | |
E. coli | |
MARSLTWRCC PWCLTEDEKA AARVDQEINR ILLEQKKQDR GELKLLLLGP GESGKSTFIK QMRIIHGAGY SEEERKGFRP LVYQNIFVSM RAMIEAMERL QIPFSRPESK HHASLVMSQD PYKVTTFEKR YAAAMQWLWR DAGIRAYYER RREFHLLDSA VYYLSHLERI TEEGYVPTAQ DVLRSRMPTT GINEYCFSVQ KTNLRIVDVG GQKSERKKWI HCFENVIALI YLASLSEYDQ CLEENNQENR MKESLALFGT ILELPWFKST SVILFLNKTD ILEEKIPTSH LATYFPSFQG PKQDAEAAKR FILDMYTRMY TGCVDGPEGS KKGARSRRLF SHYTCATDTQ NIRKVFKDVR DSVLARYLDE INLL | |
Greater than 80% as determined by SDS-PAGE. |