missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GOLM1 / GP73 Protein
A DNA sequence encoding the Homo sapiens (Human) GOLM1 / GP73, was expressed in the hosts and tags indicated.
3620.00 SEK - 20095.00 SEK
Specifications
Accession Number | NP_057632.2 |
---|---|
Gene ID (Entrez) | 51280 |
Name | GOLM1 / GP73 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036011
|
enQuireBio™
QP8697-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076011
|
enQuireBio™
QP8697-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026011
|
enQuireBio™
QP8697-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056011
|
enQuireBio™
QP8697-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066011
|
enQuireBio™
QP8697-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046011
|
enQuireBio™
QP8697-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_057632.2 | |
GOLM1 / GP73 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
51280 | |
Research Use Only | |
GOLM1 | |
Recombinant Protein | |
E. coli | |
SSRSVDLQTR IMELEGRVRR AAAERGAVEL KKNEFQGELE KQREQLDKIQ SSHNFQLESV NKLYQDEKAV LVNNITTGER LIRVLQDQLK TLQRNYGRLQ QDVLQFQKNQ TNLERKFSYD LSQCINQMKE VKEQCEERIE EVTKKGNEAV ASRDLSENND QRQQLQALSE PQPRLQAAGL PHTEVPQGKG NVLGNSKSQT PAPSSEVVLD SKRQVEKEET NEIQVVNEEP QRDRLPQEPG REQVVEDRPV GGRGFGGAGE LGQTPQVQAA LSVSQENPEM EGPERDQLVI PDGQEEEQEA AGEGRNQQKL RGEDDYNMDE NEAESETDKQ AALAGNDRNI DVFNVEDQKR DTINLLDQRE KRNHTL | |
Greater than 90% as determined by SDS-PAGE. |