missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GTPase ERas Protein
A DNA sequence encoding the Homo sapiens (Human) GTPase ERas, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8776-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_853510.1 | |
GTPase ERas Protein | |
Research Use Only | |
ERAS | |
Recombinant Protein | |
E. coli | |
LPTKPGTFDL GLATWSPSFQ GETHRAQARR RDVGRQLPEY KAVVVGASGV GKSALTIQLN HQCFVEDHDP TIQDSYWKEL TLDSGDCILN VLDTAGQAIH RALRDQCLAV CDGVLGVFAL DDPSSLIQLQ QIWATWGPHP AQPLVLVGNK CDLVTTAGDA HAAAAALAHS WGAHFVETSA KTRQGVEEAF SLLVHEIQRV QEAMAKEPMA RSCREKTRHQ KATCHCGC | |
Greater than 90% as determined by SDS-PAGE. |
3266 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |