missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Guanylate cyclase activator 2B Protein

A DNA sequence encoding the Homo sapiens (Human) Guanylate cyclase activator 2B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7537-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16009250

  • 6015.00 SEK / 100µg
Estimated Shipment: 12-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_009033.1
Guanylate cyclase activator 2B Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
2981
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Guca2b
Recombinant Protein
Yeast
VYIQYQGFRV QLESMKKLSD LEAQWAPSPR LQAQSLLPAV CHHPALPQDL QPVCASQEAS SIFKTLRTIA NDDCELCVNV ACTGCL
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers