missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HDAC8 / HDACL1 Protein

A DNA sequence encoding the Homo sapiens (Human) HDAC8 / HDACL1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6155-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061210

  • 1580.00 SEK / 10µg


Estimated Shipment
03-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_001159890.1
HDAC8 / HDACL1 Protein
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
55869
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HDAC8
Recombinant Protein
E. coli
MEEPEEPADS GQSLVPVYIY SPEYVSMCDS LAKIPKRASM VHSLIEAYAL HKQMRIVKPK VASMEEMATF HTDAYLQHLQ KVSQEGDDDH PDSIEYGLGY DCPATEGIFD YAAAIGGATI TAAQCLIDGM CKVAINWSGG WHHAKKDEAS GFCYLNDAVL GILRLRRKFE RILYVDLDLH HGDGVEDAFS FTSKVMTVSL HKFSPGFFPG TGDVSDVGLG KGRYYSVNVP IQDGIQDEKY YQICESVLKE VYQAFNPKAV VLQLGADTIA GDPMCSFNMT PVGIGKCLKY ILQWQLATLI LGGGGYNLAN TARCWTYLTG VILGKTLSSE IPDHEFFTAY GPDYVLEITP SCRPDRNEPH RIQQILNYIK GNLKHVV
Videos
SDS
Documents