missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heat shock 70 kDa protein 13 Protein

A DNA sequence encoding the Homo sapiens (Human) Heat shock 70 kDa protein 13, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8708-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036061

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_008879.3
Heat shock 70 kDa protein 13 Protein
Research Use Only
HSPA13
Recombinant Protein
E. coli
QQYLPLPTPK VIGIDLGTTY CSVGVFFPGT GKVKVIPDEN GHISIPSMVS FTDNDVYVGY ESVELADSNP QNTIYDAKRF IGKIFTAEEL EAEIGRYPFK VLNKNGMVEF SVTSNETITV SPEYVGSRLL LKLKEMAEAY LGMPVANAVI SVPAEFDLKQ RNSTIEAANL AGLKILRVIN EPTAAAMAYG LHKADVFHVL VIDLGGGTLD VSLLNKQGGM FLTRAMSGNN KLGGQDFNQR LLQYLYKQIY QTYGFVPSRK EEIHRLRQAV EMVKLNLTLH QSAQLSVLLT VEEQDRKEPH SSDTELPKDK LSSADDHRVN SGFGRGLSDK KSGESQVLFE TEISRKLFDT LNEDLFQKIL VPIQQVLKEG HLEKTEIDEV VLVGGSTRIP RIRQVIQEFF GKDPNTSVDP DLAVVTGVAI QAGIDGGSWP LQVSALEIPN KHLQKTNFN
Greater than 90% as determined by SDS-PAGE.
6782
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers