missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heat shock 70 kDa protein 13 Protein
A DNA sequence encoding the Homo sapiens (Human) Heat shock 70 kDa protein 13, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8708-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_008879.3 | |
Heat shock 70 kDa protein 13 Protein | |
Research Use Only | |
HSPA13 | |
Recombinant Protein | |
E. coli | |
QQYLPLPTPK VIGIDLGTTY CSVGVFFPGT GKVKVIPDEN GHISIPSMVS FTDNDVYVGY ESVELADSNP QNTIYDAKRF IGKIFTAEEL EAEIGRYPFK VLNKNGMVEF SVTSNETITV SPEYVGSRLL LKLKEMAEAY LGMPVANAVI SVPAEFDLKQ RNSTIEAANL AGLKILRVIN EPTAAAMAYG LHKADVFHVL VIDLGGGTLD VSLLNKQGGM FLTRAMSGNN KLGGQDFNQR LLQYLYKQIY QTYGFVPSRK EEIHRLRQAV EMVKLNLTLH QSAQLSVLLT VEEQDRKEPH SSDTELPKDK LSSADDHRVN SGFGRGLSDK KSGESQVLFE TEISRKLFDT LNEDLFQKIL VPIQQVLKEG HLEKTEIDEV VLVGGSTRIP RIRQVIQEFF GKDPNTSVDP DLAVVTGVAI QAGIDGGSWP LQVSALEIPN KHLQKTNFN | |
Greater than 90% as determined by SDS-PAGE. |
6782 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |