missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Hemoglobin subunit zeta Protein

A DNA sequence encoding the Homo sapiens (Human) Hemoglobin subunit zeta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6148-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16081170

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_005323.1
Hemoglobin subunit zeta Protein
Research Use Only
HBZ
Recombinant Protein
E. coli
SLTKTERTII VSMWAKISTQ ADTIGTETLE RLFLSHPQTK TYFPHFDLHP GSAQLRAHGS KVVAAVGDAV KSIDDIGGAL SKLSELHAYI LRVDPVNFKL LSHCLLVTLA ARFPADFTAE AHAAWDKFLS VVSSVLTEKY R
Greater than 80% as determined by SDS-PAGE.
3050
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers