missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein Q, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001153145.1 |
---|---|
Gene ID (Entrez) | 10492 |
Name | Heterogeneous nuclear ribonucleoprotein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044631
|
enQuireBio™
QP8440-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064631
|
enQuireBio™
QP8440-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054631
|
enQuireBio™
QP8440-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074631
|
enQuireBio™
QP8440-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084631
|
enQuireBio™
QP8440-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094631
|
enQuireBio™
QP8440-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001153145.1 | |
Heterogeneous nuclear ribonucleoprotein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10492 | |
Research Use Only | |
NSAP1 | |
Recombinant Protein | |
E. coli | |
ATEHVNGNGT EEPMDTTSAV IHSENFQTLL DAGLPQKVAE KLDEIYVAGL VAHSDLDERA IEALKEFNED GALAVLQQFK DSDLSHVQNK SAFLCGVMKT YRQREKQGTK VADSSKGPDE AKIKALLERT GYTLDVTTGQ RKYGGPPPDS VYSGQQPSVG TEIFVGKIPR DLFEDELVPL FEKAGPIWDL | |
Greater than 90% as determined by SDS-PAGE. |