missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein H, was expressed in the hosts and tags indicated.
1640.00 SEK - 18390.00 SEK
Specifications
Accession Number | NP_001244222.1 |
---|---|
Gene ID (Entrez) | 3187 |
Name | Heterogeneous nuclear ribonucleoprotein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16034801
|
enQuireBio™
QP8472-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074801
|
enQuireBio™
QP8472-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024801
|
enQuireBio™
QP8472-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054801
|
enQuireBio™
QP8472-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064801
|
enQuireBio™
QP8472-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044801
|
enQuireBio™
QP8472-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001244222.1 | |
Heterogeneous nuclear ribonucleoprotein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3187 | |
Research Use Only | |
HNRNPH1 | |
Recombinant Protein | |
E. coli | |
MLGTEGGEGF VVKVRGLPWS CSADEVQRFF SDCKIQNGAQ GIRFIYTREG RPSGEAFVEL ESEDEVKLAL KKDRETMGHR YVEVFKSNNV EMDWVLKHTG PNSPDTANDG FVRLRGLPFG CSKEEIVQFF SGLEIVPNGI TLPVDFQGRS TGEAFVQFAS QEIAEKALKK HKERIGHRYI EIFKSSRAEV RTHYDPPRKL MAMQRPGPYD RPGAG | |
Greater than 90% as determined by SDS-PAGE. |