Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoproteins A2 / B1 Protein
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoproteins A2/B1, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6174-ye-50ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16061340
  • 2750.00 SEK / 50µg
Estimated Shipment
03-02-2021
Specifications
NP_002128.1
Heterogeneous nuclear ribonucleoproteins A2 / B1 Protein
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
3181
Greater than 80% as determined by SDS-PAGE.
Research Use Only
HNRNPA2B1
Recombinant Protein
Yeast
MEKTLETVPL ERKKREKEQF RKLFIGGLSF ETTEESLRNY YEQWGKLTDC VVMRDPASKR SRGFGFVTFS SMAEVDAAMA ARPHSIDGRV VEPKRAVARE ESGKPGAHVT VKKLFVGGIK EDTEEHHLRD YFEEYGKIDT IEIITDRQSG KKRGFGFVTF DDHDPVDKIV LQKYHTINGH NAEVRKALSR QEMQEVQSSR SGRGGNFGFG DSRGGGGNFG PGPGSNFRGG SDGYGSGRGF GDGYNGYGG