missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoproteins A2 / B1 Protein
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoproteins A2/B1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6174-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002128.1 | |
Heterogeneous nuclear ribonucleoproteins A2 / B1 Protein | |
Research Use Only | |
HNRNPA2B1 | |
Recombinant Protein | |
E. coli | |
MEKTLETVPL ERKKREKEQF RKLFIGGLSF ETTEESLRNY YEQWGKLTDC VVMRDPASKR SRGFGFVTFS SMAEVDAAMA ARPHSIDGRV VEPKRAVARE ESGKPGAHVT VKKLFVGGIK EDTEEHHLRD YFEEYGKIDT IEIITDRQSG KKRGFGFVTF DDHDPVDKIV LQKYHTINGH NAEVRKALSR QEMQEVQSSR SGRGGNFGFG DSRGGGGNFG PGPGSNFRGG SDGYGSGRGF GDGYNGYGG | |
Greater than 80% as determined by SDS-PAGE. |
3181 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |