missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone acetyltransferase KAT7 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone acetyltransferase KAT7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8459-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186084.1 | |
Histone acetyltransferase KAT7 Protein | |
Research Use Only | |
HBO1 | |
Recombinant Protein | |
E. coli | |
MPRRKRNAGS SSDGTEDSDF STDLEHTDSS ESDGTSRRSA RVTRSSARLS QSSQDSSPVR NLQSFGTEEP AYSTRRVTRS QQQPTPVTPK KYPLRQTRSS GSETEQVVDF SDRETKNTAD HDESPPRTPT GNAPSSESDI DISSPNVSHD ESIAKDMSLK DSGSDLSHRP KRRRFHESYN FNMKCPTPGC NSLGHLTGKH ERHFSISGCP LYHN | |
Greater than 90% as determined by SDS-PAGE. |
11143 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |