missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone deacetylase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone deacetylase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6150-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004955.2 | |
Histone deacetylase 1 Protein | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3065 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
HDAC1 | |
Recombinant Protein | |
E. coli | |
MAQTQGTRRK VCYYYDGDVG NYYYGQGHPM KPHRIRMTHN LLLNYGLYRK MEIYRPHKAN AEEMTKYHSD DYIKFLRSIR PDNMSEYSKQ MQRFNVGEDC PVFDGLFEFC QLSTGGSVAS AVKLNKQQTD IAVNWAGGLH HAKKSEASGF CYVNDIVLAI LELLKYHQRV LYIDIDIHHG DGVEEAFYTT DRVMTVSFHK YGEYFPGTGD LRDIGAGKGK YYAVNYPLRD GIDDESYEAI FKPVMSKVME MFQPSAVVLQ CGSDSLSGDR LGCFNLTIKG HAKCVEFVKS FNLPMLMLGG GGYTIRNVAR CWTYETAVAL DTEIPNELPY NDYFEYFGPD FKLHISPSNM TNQNTNEYLE KIKQRLFENL RMLPHAPGVQ MQAIPEDAIP EESGDEDEDD PDKRISICSS DKRIACEEEF SDSEEEGEGG RKNSSNFKKA KRVKTEDEKE KDPEEKKEVT EEEKTKEEKP EAKGVKEEVK LA |