missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone deacetylase 6 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone deacetylase 6, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001308155.1 |
---|---|
Gene ID (Entrez) | 10013 |
Name | Histone deacetylase 6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091200
|
enQuireBio™
QP6154-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011210
|
enQuireBio™
QP6154-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001210
|
enQuireBio™
QP6154-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021210
|
enQuireBio™
QP6154-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031210
|
enQuireBio™
QP6154-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041210
|
enQuireBio™
QP6154-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001308155.1 | |
Histone deacetylase 6 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
10013 | |
Research Use Only | |
HDAC6 | |
Recombinant Protein | |
E. coli | |
MMNHCNLWDS HHPEVPQRIL RIMCRLEELG LAGRCLTLTP RPATEAELLT CHSAEYVGHL RATEKMKTRE LHRESSNFDS IYICPSTFAC AQLATGAACR LVEAVLSGEV LNGAAVVRPP GHHAEQDAAC GFCFFNSVAV AARHAQTISG HALRILIVDW DVHHGNGTQH MFEDDPSVLY VSLHRYDHGT FFPMGDEGAS SQIGRAAGTG FTVNVAWNGP RMGDADYLAA WHRLVLPIAY EFNPELVLVS AGFDAARGDP LGGCQVSPEG YAHLTHLLMG LASGRIILIL EGGYNLTSIS ESMAACTRSL LGDPPPLLTL PRPPLSGALA SITETIQVHR RYWRSLRVMK VE | |
Greater than 80% as determined by SDS-PAGE. |