missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone H3.3 Protein

A DNA sequence encoding the Homo sapiens (Human) Histone H3.3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6141-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061110

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002098.1
Histone H3.3 Protein
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
3020
Greater than 90% as determined by SDS-PAGE.
Research Use Only
H3F3A
Recombinant Protein
E. coli
ARTKQTARKS TGGKAPRKQL ATKAARKSAP STGGVKKPHR YRPGTVALRE IRRYQKSTEL LIRKLPFQRL VREIAQDFKT DLRFQSAAIG ALQEASEAYL VGLFEDTNLC AIHAKRVTIM PKDIQLARRI RGERA
Videos
SDS
Documents