missing translation for 'onlineSavingsMsg'
Läs mer

enQuireBio™ Recombinant Human Histone H3.3 Protein

Produktkod. 16031110
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 16031110

Brand: enQuireBio™ QP6141ec100ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

A DNA sequence encoding the Homo sapiens (Human) Histone H3.3, was expressed in the hosts and tags indicated.

Specifications

Tillträdesnummer NP_002098.1
Gen-ID (Entrez) 3020
Namn Histone H3.3 Protein
Kvantitet 100 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol H3F3A
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag His-SUMO
Sekvens ARTKQTARKS TGGKAPRKQL ATKAARKSAP STGGVKKPHR YRPGTVALRE IRRYQKSTEL LIRKLPFQRL VREIAQDFKT DLRFQSAAIG ALQEASEAYL VGLFEDTNLC AIHAKRVTIM PKDIQLARRI RGERA
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.